Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monkeypox Virus A29L Antibody (0027), DyLight 680 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318270FR
Description
Monkeypox Virus A29L Monoclonal antibody specifically detects Monkeypox Virus A29L in Virus samples. It is validated for Western Blot, ELISA, ELISA (Matched Antibody Pair)Specifications
Monkeypox Virus A29L | |
Monoclonal | |
DyLight 680 | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, ELISA | |
0027 | |
50mM Sodium Borate | |
Recombinant Monkeypox Virus A29L Protein. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction