Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monkeypox Virus A29L Antibody (0031), DyLight 680 , Novus Biologicals™

Monkey Monoclonal Antibody
Supplier: Novus Biologicals NBP318271FR
Description
Monkeypox Virus A29L Monoclonal antibody specifically detects Monkeypox Virus A29L in Virus samples. It is validated for Western Blot, ELISASpecifications
| Monkeypox Virus A29L | |
| Monoclonal | |
| DyLight 680 | |
| Monkey | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 0031 | |
| 50mM Sodium Borate | |
| Recombinant Monkeypox Virus A29L Protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction