Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monkeypox Virus A29L Antibody (0032), DyLight 594 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318272DL594
Description
Monkeypox Virus A29L Monoclonal antibody specifically detects Monkeypox Virus A29L in Virus samples. It is validated for Western Blot, ELISASpecifications
Monkeypox Virus A29L | |
Monoclonal | |
DyLight 594 | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, ELISA | |
0032 | |
50mM Sodium Borate | |
Recombinant Monkeypox Virus A29L Protein | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction