Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Monkeypox Virus A29L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144379
Description
Monkeypox Virus A29L Polyclonal Antibody for Western Blot, ELISA
Monkeypox Virus A29L is a gene that is found in the genome of the Monkeypox virus (MPXV). MPXV is a member of the Orthopoxvirus genus, which also includes the variola virus (which causes smallpox) and the vaccinia virus (used in smallpox vaccination). The A29L gene encodes a protein that is involved in various aspects of the viral life cycle, including viral replication, assembly, and immune evasion.
Specifications
Monkeypox Virus A29L | |
Polyclonal | |
Unconjugated | |
A29L | |
Recombinant Monkeypox virus (strain Zaire-96-I-16) (MPX) A29L protein. | |
100 μL | |
Primary | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Rabbit | |
Protein G | |
RUO | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction