Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monkeypox Virus A35 Antibody (0017), DyLight 650 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318811C
Description
Monkeypox Virus A35 Monoclonal antibody specifically detects Monkeypox Virus A35 in Virus samples. It is validated for Western Blot, ELISASpecifications
Monkeypox Virus A35 | |
Monoclonal | |
DyLight 650 | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, ELISA | |
0017 | |
50mM Sodium Borate | |
This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A35 and expressed from HEK293 cells. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction