Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monkeypox Virus A5L Antibody (0075), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP318815PE
Description
Monkeypox Virus A5L Monoclonal antibody specifically detects Monkeypox Virus A5L in Virus samples. It is validated for Western Blot, ELISASpecifications
| Monkeypox Virus A5L | |
| Monoclonal | |
| PE | |
| PBS | |
| This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A5L Protein expressed from HEK293 cells. | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| 0075 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction