Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monkeypox Virus B6R Antibody (007) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32352720UL
Description
Monkeypox Virus B6R Monoclonal antibody specifically detects Monkeypox Virus B6R in Virus samples. It is validated for Western Blot, ELISA, ELISASpecifications
Monkeypox Virus B6R | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
This antibody was obtained from a rabbit immunized with purified, recombinant Monkeypox Virus B6R (Uniprot#: QNP13760.1; Tyr18-His279). | |
20 μL | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, ELISA, ELISA | |
007 | |
Western Blot 1:1000-1:5000, ELISA 1:5000-1:10000, Sandwich ELISA Capture : 1:250-1:2000 | |
Rabbit | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction