Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monkeypox Virus E8L Recombinant Rabbit Monoclonal Antibody (7Z8N6)
GREENER_CHOICE

Catalog No. FEMA551521
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551521 Supplier Invitrogen™ Supplier No. MA551521
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Monkeypox Virus E8L Recombinant Monoclonal Antibody for Western Blot, ELISA

Monkeypox virus E8L is a protein that is expressed on the surface of the monkeypox virus. It is a conserved protein that plays a crucial role in the replication and pathogenesis of the virus. Antibodies targeting E8L have been shown to neutralize the virus and prevent infection in animal models.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monkeypox Virus E8L
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 7Z8N6
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Alias MPXV E8L
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.