Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ MONNA, >98%
GREENER_CHOICESDP

Catalog No. 577050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-705-0 50 mg
57-701-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-705-0 Supplier Tocris Bioscience™ Supplier No. 5770/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent TMEM16A (Anoctamin-1) blocker

MONNA is a potent TMEM16A (Anoctamin-1) blocker (IC50 = 80 nM). Exhibits >100-fold selectivity for TMEM16A over bestrophin-1, CLC2 and CFTR. Induces vasorelaxation of rodent resistance arteries in presence or absence of chloride ions. Carbachol (Cat. No. 2810) induced contraction of bronchial myocytes is reduced by MONNA and intracellular calcium release is increased.

Chemical Identifiers

CAS 1572936-83-4
Molecular Formula C18H14N2O5

Specifications

Quantity 50 mg
Solubility Soluble to 100mM in DMSO
Formula Weight 338.31
Purity >98%
Molecular Weight (g/mol) 338.319
Chemical Name or Material 2-[(4-Methoxy-2-naphthalenyl)amino]-5-nitro-benzoic acid
Recommended Storage Store at 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.