Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mono-6-O-(p-toluenesulfonyl)-alpha-cyclodextrin 85.0+%, TCI America™
SDP

Catalog No. M16441G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M16441G 1 g
M1644200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M16441G Supplier TCI America Supplier No. M16441G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32860-56-3
Molecular Formula C43H66O32S
Molecular Weight (g/mol) 1127.029
InChI Key ARQITQMHQNGIEE-FJFUKXEISA-N
PubChem CID 44629832
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2C3C(C(C(O2)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O3)C(C8O)O)CO)CO)CO)CO)CO)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 1127.03
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mono-6-O-(p-toluenesulfonyl)-alpha-cyclodextrin
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.