Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mono-Methyl Arginine Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116819
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116819 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116819 Supplier Invitrogen™ Supplier No. PA5116819
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HCT116, Mouse lung, Mouse liver, Mouse brain, Rat brain.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mono-Methyl Arginine
Applications Dot Blot, ELISA, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Alias monomethyl arginine
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence containing monomethylated R.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical, Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.