Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Mono-methyl-HIST1H2AG (Lys9) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144299
Description
Mono-methyl-HIST1H2AG (Lys9) Polyclonal Antibody for ICC/IF, ChIP, ELISA
Histones are nuclear proteins that play an important role in maintaining the structure of the chromosome in eukaryotes. The nucleosome is made up of around 146bp of DNA, which enfolds around a histone octomer unit comprising of four major histones, H2A, H2B, H3 and H4. An important role of H2A is to ensure proper assembly or functioning of the centrometre. They also play an important role in gene regulation. The protein can be modified by acetylation and phosphorylation.
Specifications
Mono-methyl-HIST1H2AG (Lys9) | |
Polyclonal | |
Unconjugated | |
H2AC11 | |
H2A histone family, member P; H2A.1; H2A.1b; H2A/d; H2A/p; H2AC11; H2AC13; H2AC15; H2AC16; H2AC17; H2AFC; H2AFD; H2AFI; H2AFN; H2AFP; H2AG; H2ak; HIST1H2AG; HIST1H2AI; HIST1H2AJ; HIST1H2AK; HIST1H2AL; HIST1H2AM; histone 1, H2ag; histone cluster 1, H2ag; histone H2A type 1; histone H2A/e; histone H2A/p; Histone H2A/ptl; pH2A/f | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8329, 8969 | |
-20°C or -80°C if preferred | |
Liquid |
ChIP Assay, ELISA, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P0C0S8 | |
H2AC11, HIST1H2AG | |
Peptide sequence around site of Mono-methyl-Lys (9) derived from Human Histone H2A type 1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction