Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mono-methyl-HIST1H2BC (Lys15) Polyclonal Antibody

Mono-methyl-HIST1H2BC (Lys15) Polyclonal Antibody
Supplier: Thermo Scientific PA5144329
Description
Mono-methyl-HIST1H2BC (Lys15) Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Specifications
Mono-methyl-HIST1H2BC (Lys15) | |
Polyclonal | |
Unconjugated | |
H2BC5 | |
dJ221C16.3; H2B Clustered Histone 4; H2B histone family, member L; H2B.1; H2B/a; H2B/g; H2B/h; H2B/k; H2B/l; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFG; H2BFH; H2BFK; H2BFL; HIST1H2BC; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BI; histone 1, H2bc; histone cluster 1, H2bc; histone H2B type 1-C/E/F/G/I; Histone H2B.1 A; Histone H2B.a; Histone H2B.g; Histone H2B.h; Histone H2B.k; Histone H2B.l | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3017 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
P58876, P62807 | |
H2BC5, Hist1h2bc | |
Peptide sequence around site of Mono-methyl-Lys (15) derived from Human Histone H2B type 1-C/E/F/G/I. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction