Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mono-methyl phthalate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
4376-18-5
C9H8O4
180.16
MFCD00002466
FNJSWIPFHMKRAT-UHFFFAOYSA-N
2-methoxycarbonyl benzoic acid,methyl hydrogen phthalate,monomethyl phthalate,mono-methyl phthalate,phthalic acid monomethyl ester,o-methoxycarbonyl benzoic acid,monomethyl 1,2-benzenedicarboxylate,phthalic acid, monomethyl ester,monomethylphthalate,unii-2stt6d18jr
20392
2-methoxycarbonylbenzoic acid
COC(=O)C1=CC=CC=C1C(=O)O
4376-18-5
C9H8O4
180.16
MFCD00002466
FNJSWIPFHMKRAT-UHFFFAOYSA-N
2-methoxycarbonyl benzoic acid,methyl hydrogen phthalate,monomethyl phthalate,mono-methyl phthalate,phthalic acid monomethyl ester,o-methoxycarbonyl benzoic acid,monomethyl 1,2-benzenedicarboxylate,phthalic acid, monomethyl ester,monomethylphthalate,unii-2stt6d18jr
20392
2-methoxycarbonylbenzoic acid
COC(=O)C1=CC=CC=C1C(=O)O
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 81.0°C to 86.0°C
Color White
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (COOH)C6H4COOCH3
Solubility Information Solubility in water: insoluble (20°C). Other solubilities: soluble in ethanol and ether
Formula Weight 180.16
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material mono-Methyl phthalate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-476-6