Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monoacylglycerol Lipase Monoclonal Antibody (OTI1B1)
GREENER_CHOICE

Catalog No. PIMA525582
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525582 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525582 Supplier Invitrogen™ Supplier No. MA525582
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoacylglycerol Lipase Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

Monoglyceride lipase functions together with hormone-sensitive lipase to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monoacylglycerol Lipase
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI1B1
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MGLL
Gene Accession No. Q99685
Gene Alias AA589436; HUK5; HU-K5; lysophospholipase homolog; Lysophospholipase-like; MAGL; Mgl; Mgl2; MGLL; Monoacylglycerol lipase; monoglyceride lipase
Gene Symbols MGLL
Host Species Mouse
Immunogen Full length human recombinant protein of MGLL produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11343
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.