Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monoacylglycerol Lipase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552891
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552891 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552891 Supplier Invitrogen™ Supplier No. PA552891
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RQHIMETGPE DPSSMPEESS PRRTPQSIPY QDLPHLVNAD GQYLFCRYWK PTGTPKALIF VSHGAGEHSG RYEELARMLM GLDLLVFAHD HVGHGQSEGE RMVVSDFHVF VRDVLQHVDS MQKDYPGLPV FLLGHSM Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 82%.

Monoglyceride lipase functions together with hormone-sensitive lipase to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monoacylglycerol Lipase
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MGLL
Gene Accession No. Q99685
Gene Alias AA589436; HUK5; HU-K5; lysophospholipase homolog; Lysophospholipase-like; MAGL; Mgl; Mgl2; MGLL; Monoacylglycerol lipase; monoglyceride lipase
Gene Symbols MGLL
Host Species Rabbit
Immunogen Recombinant protein fragment of Human MGLL (Product #RP-88953)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.