Learn More
Invitrogen™ Monoamine Oxidase B Polyclonal Antibody

Description
Recommended positive controls: HepG2, mouse liver. Predicted reactivity: Mouse (87%), Rat (86%), Dog (94%), Pig (90%), Bovine (90%), Guinea pig (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Monoamine Oxidase B |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.51 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | MAOB |
| Gene Accession No. | P19643, P27338, Q8BW75 |
| Gene Alias | 6330414K01Rik; adrenalin oxidase; amine oxidase [flavin-containing] B; HGNC:6834; MAO b; MAO, brain; MAO, platelet; Maob; MAO-B; MGC26382; monoamine oxidase B; monoamine oxidase type B; monoamine oxidase-B; RP1-201D17__B.1; tyramine oxidase |
| Gene Symbols | MAOB |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.