Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monobasic Sodium Phosphate, Dihydrate, USP, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18604664 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
500 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

S1930, 13472-35-0, NaH2PO4·2H2O

Spectrum™ Chemical Monobasic Sodium Phosphate, Dihydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13472-35-0
Molecular Formula H6NaO6P
Molecular Weight (g/mol) 156.01
InChI Key VBJGJHBYWREJQD-UHFFFAOYSA-M
IUPAC Name sodium dihydrate dihydrogen phosphate
SMILES O.O.[Na+].OP(O)([O-])=O

Specifications

pH 4.1 to 4.5
CAS Max % 1
Quantity 12 kg
Percent Purity 98 to 103%
Grade USP
Packaging Poly Pail
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.