Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Monocytes/Granulocytes Monoclonal Antibody (74-22-15), PE
GREENER_CHOICE

Catalog No. PIMA528824
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528824 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528824 Supplier Invitrogen™ Supplier No. MA528824
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monocytes/Granulocytes Monoclonal Antibody for Flow

Monocyte/granulocyte antibody reacts with > 90% of porcine monocytes and granulocytes and < 5% of peripheral blood lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Monocytes/Granulocytes
Applications Flow Cytometry
Classification Monoclonal
Clone 74-22-15
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.5
Gene Alias CD172a; SWC3a
Host Species Mouse
Immunogen Fresh dd miniature swine thymocytes.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.