Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethyl Pimelate 98.0+%, TCI America™
SDP

Catalog No. P124425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P124425G 25 g
P12445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P124425G Supplier TCI America Supplier No. P124425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33018-91-6
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00801202
InChI Key NQYXFXWKKYGBNL-UHFFFAOYSA-N
Synonym ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester
PubChem CID 118383
IUPAC Name 7-ethoxy-7-oxoheptanoic acid
SMILES CCOC(=O)CCCCCC(=O)O

Specifications

Color Yellow
Boiling Point 162°C
Quantity 25 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Monoethyl Pimelate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.