Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monoglyceride Lipase Antibody (HL1937) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325378
Description
Monoglyceride Lipase Monoclonal antibody specifically detects Monoglyceride Lipase in Human samples. It is validated for Western BlotSpecifications
Monoglyceride Lipase | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
HL1937 | |
Western Blot 1:500-1:3000 | |
HU-K5EC 3.1.1.23, Lysophospholipase homolog, Lysophospholipase-like, MAGL, MGLHUK5, Monoacylglycerol lipase, monoglyceride lipase | |
Recombinant fragment of human Monoglyceride Lipase. | |
100 μL | |
Lipid and Metabolism | |
11343 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction