Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Monoglyceride Lipase Antibody (OTI2B11), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272759MFV450
Description
Monoglyceride Lipase Monoclonal antibody specifically detects Monoglyceride Lipase in Human, Monkey samples. It is validated for Flow CytometrySpecifications
Monoglyceride Lipase | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Monkey | |
Purified |
Flow Cytometry | |
OTI2B11 | |
Flow Cytometry | |
HU-K5EC 3.1.1.23, Lysophospholipase homolog, Lysophospholipase-like, MAGL, MGLHUK5, Monoacylglycerol lipase, monoglyceride lipase | |
Full length human recombinant protein of human MGLL (NP_009214) produced in HEK293T cell. | |
0.1 mL | |
Lipid and Metabolism | |
11343 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction