Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Covidien Monoject™ Tube Caps

Catalog No. 0254424
Change view
Click to view available options
Color:
Blue
Green
Lavender
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
02-544-24 Green
02-544-27 White
02-544-26 Blue
02-544-25 Lavender
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 02-544-24 Supplier Covidien Supplier No. 1270019503
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Easy-on, easy-off caps for glass blood collection tubes

  • Exclusive thumb tab makes cap removal simple, quick and fatigue-free
  • Supplied in convenient, resealable and polybag-lined dispenser box
  • Ideal for benchtop use
  • Available in six colors to match the original blood tube stopper
  • Size: 13mm

Specifications

Color Green
For Use With (Equipment) 13 mm glass blood collection tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.