Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomagnesium EDTA, 0.2 g/L, for Hardness, Ricca Chemical
SDP

Catalog No. 521232 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 14402-88-1, 7732-18-5
Molecular Formula C10H12MgN2Na2O8
Molecular Weight (g/mol) 358.50
MDL Number MFCD00078217
InChI Key AWNVVAMWLMUZOZ-UHFFFAOYSA-J
Synonym unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate
PubChem CID 161064
IUPAC Name magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless
CAS Min % 98.0, 0.02
CAS Max % 100.0, 0.02
Grade Laboratory
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Concentration or Composition Notes 0.2g/L
Chemical Name or Material Monomagnesium EDTA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.