Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate 98.0+%, TCI America™
SDP

Catalog No. M249325G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Catalog No. M249325G

Supplier: TCI America M249325G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 74936-72-4
Molecular Formula C16H16N2O6
Molecular Weight (g/mol) 332.312
MDL Number MFCD00475478
InChI Key JPXPPUOCSLMCHK-UHFFFAOYSA-N
Synonym 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid Monomethyl Ester
PubChem CID 173322
IUPAC Name 5-methoxycarbonyl-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3-carboxylic acid
SMILES CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC(=CC=C2)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 25 g
Formula Weight 332.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.