Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ monomethyl Histone H2A (Gln104), Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE1332MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE1332MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE1332MI Supplier MilliporeSigma™ Supplier No. ABE1332
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H2A type 1 (UniProt P0C0S8; also known as H2A.1, Histone H2A/p) is encoded by the HIST1H2AG (also known as H2AFP, HIST1H2AI, H2AFC, HIST1H2AK, H2AFD, HIST1H2AL, H2AFI, HIST1H2AM, H2AFN) gene (Gene ID 8969) in human. Histone H2A is one of four components of the core nucleosome structure. The nucleosome represents a unit of chromatin in which DNA is wrapped around a histone octamer. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones. Histones undergo a number of post-translational modifications in response to various stimuli that may induce changes in the structure of the nucleosome and hide or expose DNA sequences. Histone H2A, in particular, undergoes acetylation on lysine 5 by Tip60 enzyme, which may promote unfolding of chromatin and transcription. In addition, H2A is phosphorylated on serine 1 and serine 139 (H2A.X) residues, which mark mitosis and gene repression and DNA damage, respectively. Gene repression may also result from the sumoylation of H2A s lysine 126 residue. In addition, ubiquitination of the H2A lysine 119 residue may play a role in spermatogenesis. Monoubiquitination of Lys-120 (H2AK119Ub) by RING1, TRIM27 and RNF2/RING2 complex gives a specific tag for epigenetic transcriptional repression and participates in X chromosome inactivation of female mammals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen monomethyl Histone H2A (Gln104)
Applications Dot Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene HIST1H2AG, H2AFP, HIST1H2AI, H2AFC, HIST1H2AK, H2AFD, HIST1H2AL, H2AFI, HIST1H2AM, H2AFN
Gene Accession No. P0C0S8
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human histone H2A target region sequence containing monomethylated Gln104 (Q104me).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_003500
Test Specificity Specificity of this polyclonal antibody was verified by dot blot analysis. This polyclonal antibody specifically detected peptide corresponding to histone H2A target region sequence with monomethylated Gln104 (Q104me), but not H2A peptides without Q104me modification, nor peptides derived from histone H1, H2A.Z, H2B, H3, H3.3, or H4 sequences.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.