Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
monomethyl Histone H2B (Pro1) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $525.84
Specifications
Antigen | monomethyl Histone H2B (Pro1) |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Inhibition Assays |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE1464-25UG
|
MilliporeSigma
ABE146425UG |
25 μg |
Each for $192.35
|
|
|||||
ABE1464-100
|
MilliporeSigma
ABE1464100UG |
100 μg |
Each for $525.84
|
|
|||||
Description
Anti-monomethyl Histone H2B (Pro1) Antibody, Cat. No. ABE1464, is a highly specific rabbit polyclonal antibody that targets Histone H2B monomethylated on proline 1 and has been tested for use in Western Blotting and Peptide Inhibition Assay.Specifications
monomethyl Histone H2B (Pro1) | |
Western Blot, Inhibition Assays | |
Unconjugated | |
Rabbit | |
Epigenetics & Nuclear Function | |
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
NP_003509 | |
KLH-conjugated linear peptide corresponding to the first 9 amino acids (methiionine removed) from the N-terminus region of human Histone H2B1C with methylated proline. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Histone H2B type 1-C/E/F/G/I;Histone H2B.1 A;Histone H2B.a;H2B/a;Histone H2B.g;H2B/g;Histone H2B.h;H2B/h;Histone H2B.k;H2B/k;Histone H2B.l;H2B/l | |
HIST1H2BC;H2BFL;HIST1H2BE;H2BFH;HIST1H2BF;H2BFG;HIST1H2BG;H2BFA;HIST1H2BI;H2BFK | |
IgG | |
Affinity Purified | |
This rabbit polyclonal antibody specifically detect Histone H2B monomethylated on proline 1. | |
∽16 kDa observed; 13.91 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title