Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Montmorillonite K-10, granules, Acros Organics

Click to view available options
Quantity:
100g
1kg
5kg
1318-93-0
Al2H2O12Si4
360.307
GUJOJGAPFQRJSV-UHFFFAOYSA-N
montmorillonite,bentolite,gelwhite l,montmorillonite ksf,montmorillonite k 10,unii-a585mn1h2l,envirobent,neokunibond,arcillite,bedelix
71586775
dialuminum;dioxosilane;oxygen(2-);hydrate
O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]
1318-93-0
Al2H2O12Si4
360.307
GUJOJGAPFQRJSV-UHFFFAOYSA-N
montmorillonite,bentolite,gelwhite l,montmorillonite ksf,montmorillonite k 10,unii-a585mn1h2l,envirobent,neokunibond,arcillite,bedelix
71586775
dialuminum;dioxosilane;oxygen(2-);hydrate
O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]
Acidity 2.5 mg KOH/g (free acidity) max. 10 to 14 mg KOH/g (total acidity)
Fieser 15,213
Merck Index 11,6168
Density 0.500 to 0.600 kg/l
Particle Size 10% max. (< 0.5 mm) 80% min. (0.5−1,25 mm) 10% max. (> 1.25 mm)
Quantity 100g
Chemical Name or Material Montmorillonite K-10

EINECSNumber : 215-288-5