Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Montmorillonite K-10, granules

Catalog No. p-7077599
Click to view available options
Quantity:
100 g

CAS: 1318-93-0 | Al2H2O12Si4 | 360.307 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1318-93-0
Molecular Formula Al2H2O12Si4
Molecular Weight (g/mol) 360.307
InChI Key GUJOJGAPFQRJSV-UHFFFAOYSA-N
Synonym montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
PubChem CID 71586775
IUPAC Name dialuminum;dioxosilane;oxygen(2-);hydrate
SMILES O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]

Specifications

Quantity 100 g
Fieser 15,213
Merck Index 11,6168
Density 0.500 to 0.600 kg/l
Chemical Name or Material Montmorillonite K-10
EINECSNumber 215-288-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.