Learn More
Moore Medical Apothecary Products™ Eye Patch
Description
- Elastic band holds patch securely
- Soft, black fabric
Specifications
Specifications
| For Use With (Application) | Comfort and protection |
| Product Type | Eye Patch |
| Color | Black |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.