Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOPS-SDS Running Buffer, 20X, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
Description
General Description
- MOPS-SDS running buffer is a zwitterionic, morpholinic buffer that allows the separation of medium- to large-sized proteins
Application
- MOPS is used as a running buffer in electrophoresis. The use of MOPS buffer allows proteins to run slower than when MES buffer is used
- MOPS-SDS running buffer, 20X concentrated, is formulated to be used for running proteins on an SDS gel
- This buffer contains SDS detergent to maintain the denaturation of proteins and MOPS to buffer the pH
- It is commonly used for cell culture media and protein purification in chromatography
- MOPS is frequently applied in buffered culture media for bacteria, yeast, and mammalian cells
- It is also applied in the bicinchoninic acid (BCA) assay
- It is used in separating RNA in agarose gels

Specifications
Specifications
Chemical Name or Material | MOPS-SDS running buffer |
Concentration | 20X |
Physical Form | Liquid |
Quantity | 1 L |
Solubility Information | It is soluble in water. |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction