Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOR26-1,2,3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP280542
Description
MOR26-1,2,3 Polyclonal specifically detects MOR26-1,2,3 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MOR26-1,2,3 | |
Polyclonal | |
Western Blot 1:300 - 1:2000, Immunohistochemistry 1:300 - 1:2000, Immunohistochemistry-Paraffin 1:300 - 1:2000 | |
Olfactory Receptor 52P1, Olfr608, Olfr622, Olfr629, OR11-66, OR52P1 | |
A synthetic peptide from mouse MOR26-1,2,3 conjugated to blue carrier protein was used as the antigen. | |
0.1 mL | |
Primary | |
Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Lyophilized from whole serum | |
Rabbit | |
Unpurified | |
RUO | |
259087 | |
Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction