Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MORC2 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600236G
Description
MORC2 Polyclonal antibody specifically detects MORC2 in Human samples. It is validated for Western BlotSpecifications
MORC2 | |
Polyclonal | |
Western Blot | |
AC004542.C22.1, KIAA0852, MORC family CW-type zinc finger 2, MORC family CW-type zinc finger protein 2, ZCW3, ZCWCC1, Zinc finger CW-type coiled-coil domain protein 1, zinc finger, CW type with coiled-coil domain 1, zinc finger, CW-type with coiled-coil domain 1 | |
A synthetic peptide, which maps to the region between residues 600-700 of human zinc finger, CW-type with coiled-coil domain 1 using the numbering given in entry NP_055756.1 (GeneID 22880). | |
0.1 mL | |
Transcription Factors and Regulators | |
22880 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction