Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551390
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551390 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551390 Supplier Invitrogen™ Supplier No. PA551390
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSECLRIEPD TTALSTNHET IDLLVQILRN CLRYFLPPSF PISKKQLSAM NSDELISFPL KEYFKQYEVG LQNLCNSYQS RADSRAKASE ESLRTSERKL RETEEKLQKL RTNIVALLQK VQEDIDINTD DELDAYIED Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.

Exhibits a cytosolic function in lipogenesis, adipogenic differentiation, and lipid homeostasis by increasing the activity of ACLY, possibly preventing its dephosphorylation. May act as a transcriptional repressor. Down-regulates CA9 expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORC2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MORC2
Gene Accession No. Q9Y6X9
Gene Alias 8430403M08Rik; ATPase MORC2; ATPase MORC2A; CMT2Z; Kiaa0852; microrchidia 2A; MORC family CW-type zinc finger 2; MORC family CW-type zinc finger protein 2; MORC family CW-type zinc finger protein 2A; MORC2; Morc2a; ZCW3; ZCWCC1; zinc finger CW-type coiled-coil domain protein 1; zinc finger, CW-type with coiled-coil domain 1
Gene Symbols MORC2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MORC2. Recombinant protein control fragment (Product #RP-91359).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22880
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.