Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MORC3 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306255AF647
Description
MORC3 Polyclonal specifically detects MORC3 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
MORC3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
KIAA0136, MORC family CW-type zinc finger 3, MORC family CW-type zinc finger protein 3, nuclear matrix protein NXP2, NXP2, ZCW5, ZCWCC3, Zinc finger CW-type coiled-coil domain protein 3, zinc finger, CW type with coiled-coil domain 3, zinc finger, CW-type with coiled-coil domain 3 | |
Produced in rabbits immunized with E. coli-derived Human MORC3 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
23515 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction