Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MORC3 Mouse anti-Human,Mouse,Rat, Unconjugated, Clone: 17A9, MilliporeSigma™

Catalog No. MABC11051MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC11051MI 100 μg
MABC11052MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC11051MI Supplier MilliporeSigma Supplier No. MABC1105100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MORC3 Monoclonal antibody specifically detects MORC3 in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORC3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 17A9
Conjugate Unconjugated
Gene Accession No. NP_056173
Gene Alias MORC family CW-type zinc finger protein 3;Nuclear matrix protein 2;Zinc finger CW-type coiled-coil domain protein 3
Gene Symbols MORC3, KIAA0136, NXP2, ZCWCC3
Host Species Mouse
Immunogen His-tagged full-length recombinant human MORC family CW-type zinc finger protein 3 (MORC3).
Molecular Weight of Antigen ∼160 and 230 kDa observed; 107.11 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 17A9 is a mouse monoclonal antibody that specifically detects MORC family CW-type zinc finger protein 3 (MORC3). It targets an epitope with the C-terminal region.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.