Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORC3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542786 Supplier Invitrogen™ Supplier No. PA542786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KLLAELDAII GKKGTRIIIW NLRSYKNATE FDFEKDKYDI RIPEDLDEIT Sequence homology: Cow: 86%; Dog: 86%; Guinea Pig: 100%; Horse: 79%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 93%.

This gene encodes a protein that localizes to the nuclear matrix. The protein also has RNA binding activity, and has a predicted coiled coil domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORC3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MORC3
Gene Accession No. Q14149
Gene Alias 1110051N18Rik; AI452146; BF318192; D16Jhu32e; KIAA0136; microrchidia 3; MORC family CW-type zinc finger 3; MORC family CW-type zinc finger protein 3; Morc3; Nuclear matrix protein 2; nuclear matrix protein NXP2; NXP2; ZCW5; Zcwcc3; Zinc finger CW-type coiled-coil domain protein 3; zinc finger, CW type with coiled-coil domain 3; zinc finger, CW-type with coiled-coil domain 3
Gene Symbols MORC3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MORC3 (aa 180-229).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23515
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.