Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MORC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111045
Description
Immunogen sequence: GKKGTRIIIW NLRSYKNATE FDFEKDKYDI RIPEDLDEIT GKKGYKKQER MDQIAPESDY SLRAYCSILY LKPRMQIILR.
This gene encodes a protein that localizes to the nuclear matrix. The protein also has RNA binding activity, and has a predicted coiled coil domain.
Specifications
| MORC3 | |
| Polyclonal | |
| Unconjugated | |
| MORC3 | |
| 1110051N18Rik; AI452146; BF318192; D16Jhu32e; KIAA0136; microrchidia 3; MORC family CW-type zinc finger 3; MORC family CW-type zinc finger protein 3; Morc3; Nuclear matrix protein 2; nuclear matrix protein NXP2; NXP2; ZCW5; Zcwcc3; Zinc finger CW-type coiled-coil domain protein 3; zinc finger, CW type with coiled-coil domain 3; zinc finger, CW-type with coiled-coil domain 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23515 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14149 | |
| MORC3 | |
| Recombinant Protein corresonding to Human MORC3. Recombinant protein control fragment (Product #RP-105889). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction