Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morc3 Polyclonal Antibody, Rockland™
SDP

Catalog No. 501990148
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0148 25 μL
50-199-0147 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-0148 Supplier Rockland Immunochemicals Supplier No. 100401N97S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. The antibody is specific for human Morc3 in expressed cell lysates. Cross reactivity is seen in mouse Morc3. Cross reactivity to other Morc proteins has not been determined.

This gene encodes a protein that localizes to the nuclear matrix. The protein also has RNA binding activity, and has a predicted coiled coil domain.

Specifications

Antigen Morc3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 70 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene MORC3
Gene Accession No. F7BJB9, Q14149
Gene Alias 1110051N18Rik; AI452146; BF318192; BOS_1467; D16Jhu32e; KIAA0136; LOC100348526; microrchidia 3; MORC family CW-type zinc finger 3; MORC family CW-type zinc finger protein 3; Morc3; Nuclear matrix protein 2; nuclear matrix protein NXP2; Nxp2; ZCW5; ZCWCC3; Zinc finger CW-type coiled-coil domain protein 3; zinc finger, CW type with coiled-coil domain 3; zinc finger, CW-type with coiled-coil domain 3
Gene Symbols MORC3
Host Species Rabbit
Immunogen Morc3 whole rabbit serum was prepared by repeated immunizations with a human Morc3 recombinant protein.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23515, 338467
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.