Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORF Monoclonal Antibody (8C8D12)
GREENER_CHOICE

Catalog No. PIMA538548
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538548 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538548 Supplier Invitrogen™ Supplier No. MA538548
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody has been tested in indirect ELISA.

MOZ (monocytic leukemia zinc finger protein) is a chromatin-associated histone acetyltransferase (HAT) that regulates chromatin remodeling and transcription. The MOZ gene was initially isolated as a consequence of two variant translocations that were identified in a distinct subtype of acute myeloid leukemias and resulted in the formation of MOZ fusion proteins. These fusions involve the HAT domain of MOZ with the activation domain of either transcriptional coactivator protein TIF2/GRIP1 or CBP, and lead to enhanced transcriptional activation by a mechanism involving aberrant histone acetylation. Additional MOZ related proteins, including MORF (MOZ related factor) and Tip60 (TAT interacting proteins 60), share significant similarities with MOZ including the putuative HAT domain. MORF also contains a strong transcriptional repression domain at its N terminus and a highly potent activation domain at the C terminus, suggesting that MORF has both HAT activity and contributes to the regulation of transcriptional activation. Tip60 was originally identified as a coactivator for the HIV TAT protein and also functions as a nuclear hormone receptor coactivator that enhances ligand dependent steroid receptor-mediated transactivation involving the androgen, estrogen and progesterone receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORF
Applications ELISA, Western Blot
Classification Monoclonal
Clone 8C8D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene KAT6B
Gene Accession No. Q8WYB5
Gene Alias AI507552; B130044K16Rik; GTPTS; histone acetyltransferase KAT6B; histone acetyltransferase MORF; histone acetyltransferase MOZ2; histone acetyltransferase MYST4; histone acetyltransferase querkopf; K(lysine) acetyltransferase 6B; Kat6b; KIAA0383; lysine acetyltransferase 6B; mKIAA0383; monocytic leukemia; monocytic leukemia zinc finger protein-related factor; MORF; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 4; MOZ2; MOZ-related factor; MYST histone acetyltransferase (monocytic leukemia) 4; MYST histone acetyltransferase monocytic leukemia 4; Myst4; MYST-4; protein querkopf; qkf; querkopf; SAS2; TIP60 protein 4; ZC2HC6B
Gene Symbols KAT6B
Host Species Mouse
Immunogen Purified recombinant fragment of human KAT6B (aa: 1186-1318) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.