Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MORF4 Antibody (OTI5F2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272769F
Description
MORF4 Monoclonal specifically detects MORF4 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry.Specifications
MORF4 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
MORF4 | |
Recombinant protein fragment corresponding to amino acids 1-255 of human MORF4 (NP_006783) produced in E.coli. | |
0.1 mL | |
10934.0 | |
Store at 4°C in the dark. |
Immunohistochemistry | |
OTI5F2 | |
Immunohistochemistry | |
CSR, CSRB, mortality factor 4, SEN, SEN1, senescence (cellular)-related 1, senescence-related, cellular, 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction