Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORF4 Monoclonal Antibody (OTI5F2)
GREENER_CHOICE

Catalog No. PIMA526430
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526430 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526430 Supplier Invitrogen™ Supplier No. MA526430
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MORF4 Monoclonal Antibody for Western Blot, IHC (P)

Cellular senescence is the terminal non-dividing state that normal cells enter following completion of their proliferative potential. Fusions of immortal human cell lines with each other have led to their assignment to one of four complementation groups. Mortality factor 4 (MORF4) was identified as the lead member of a family of transcription factor-like proteins that reverses this immortal phenotype. Like other members in this family, MORF4 is localized to the nucleus and possesses transcription factor-like motifs such as helix-loop-helix and a leucine zipper motif that might allow it to form transcriptionally active homo- or heterodimers. MORF4 has been shown to bind to the transcription corepressors mSin3A and TLE, suggesting that together, these complexes may play a role in transcriptional repression of genes that lead to cellular senescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORF4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI5F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MORF4
Gene Accession No. 0
Gene Alias 9130413I22Rik; AL022827; csrb; Eaf3; Esa1p-associated factor 3 homolog; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; mKIAA4002; MNCb-1039; Mof4 family associated protein 1; Morf4 family associated protein 1; MORF4 family-associated protein 1; MORF4L1; MORF-related gene 15 protein; MORF-related gene on chromosome 15; MORFRG15; mortality factor 4 like 1; mortality factor 4-like protein 1; Mrfap1; mrfap1 {ECO:0000312; MRG15; Pam14; Pgr1; PP368; Protein associated with MRG of 14 kDa; protein associated with MRG, 14 kDa; protein MSL3-1; RGD:628631}; RGD1306635; S863-6; SEN1; T-cell activation protein; T-cell activation protein-related protein; TEG-189; testis expressed gene 189; testis-expressed gene 189 protein; Tex189; transcription factor-like protein MRG15
Gene Symbols MORF4
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-255 of MORF4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10934
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.