Learn More
Invitrogen™ MORF4 Polyclonal Antibody

Description
A suggested positive control is K562 cell lysate. PA5-20399 can be used with blocking peptide PEP-0516.
Specifications
Specifications
| Antigen | MORF4 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | MORF4 |
| Gene Accession No. | 0 |
| Gene Alias | 9130413I22Rik; AL022827; csrb; Eaf3; Esa1p-associated factor 3 homolog; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; mKIAA4002; MNCb-1039; Mof4 family associated protein 1; Morf4 family associated protein 1; MORF4 family-associated protein 1; MORF4L1; MORF-related gene 15 protein; MORF-related gene on chromosome 15; MORFRG15; mortality factor 4 like 1; mortality factor 4-like protein 1; Mrfap1; mrfap1 {ECO:0000312; MRG15; Pam14; Pgr1; PP368; Protein associated with MRG of 14 kDa; protein associated with MRG, 14 kDa; protein MSL3-1; RGD:628631}; RGD1306635; S863-6; SEN1; T-cell activation protein; T-cell activation protein-related protein; TEG-189; testis expressed gene 189; testis-expressed gene 189 protein; Tex189; transcription factor-like protein MRG15 |
| Gene Symbols | MORF4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.