Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MORF4L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596568
Description
Immunogen sequence: MAPKQDPKPK FQEGERVLCF HGPLLYEAKC VKVAIKDKQV KYFIHYSGWN KNWDEWVPES RVLKYVDTNL QKQRELQKAN QEQYAEGKMR GAAPGKKTSG; Positive Samples: MCF-7, HepG2, THP-1, Mouse brain, Mouse spleen, Mouse testis, Rat brain; Cellular Location: Nucleus.
MRG15 ia a component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome-DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis and DNA repair.
Specifications
MORF4L1 | |
Polyclonal | |
Unconjugated | |
MORF4L1 | |
Eaf3; Esa1p-associated factor 3 homolog; FWP006; HSPC008; HSPC061; HsT17725; MEAF3; MGC10631; mKIAA4002; Morf4l1; MORF-related gene 15 protein; MORF-related gene on chromosome 15; MORFRG15; mortality factor 4 like 1; mortality factor 4-like protein 1; MRG15; PP368; Protein MSL3-1; S863-6; TEG-189; testis expressed gene 189; testis-expressed gene 189 protein; Tex189; transcription factor-like protein MRG15 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10933, 21761, 300891 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.71 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60762, Q6AYU1, Q9UBU8 | |
MORF4L1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human MORF4L1 (NP_0067821). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction