Learn More
Invitrogen™ MORF4L2 Polyclonal Antibody

Description
Positive Samples: Cellular Location: Nucleus Immunogen sequence: MSSRKQGSQP RGQQSAEEEN FKKPTRSNMQ RSKMRGASSG KKTAGPQQKN LEPALPGRWG GRSAENPPSG SVRKTRKNKQ KTPGNGDGGS TSEAPQPPRK KRARADPTVE SEEAFKNRME VKVKIPEELK PWLVEDWDLV TRQKQLFQLP AKKNVDAILE EYANCKKSQG NVDNKEYAVN EVVAGIKEYF NVMLGTQLLY KFERPQYAEI LLAHPDAPMS QVYGAPHLLR LFVRIGAMLA YTPLDEKSLA LLLGYLHDFL KYLAKNSASL FTASDYKVAS AE.
Specifications
Specifications
| Antigen | MORF4L2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.66 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MORF4L2 |
| Gene Accession No. | Q15014, Q6QI89, Q9R0Q4 |
| Gene Alias | 2410017O14Rik; Kiaa0026; Liver regeneration-related protein LRRG00119; LRRG00119; mKIAA0026; Morf4l2; MORFL2; MORF-related gene X; MORF-related gene X protein; mortality factor 4 like 2; mortality factor 4-like protein 2; MRGX; MSL3-2 protein; OTTHUMP00000023756; protein MSL3-2; RP5-1055C14.2; Sid 393; Sid393; Sid393p; transcription factor-like protein Mrgx |
| Gene Symbols | MORF4L2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.