Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MORF4L2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540765
Description
Peptide sequence: MSSRKQGSQP RGQQSAEEEN FKKPTRSNMQ RSKMRGASSG KKTAGPQQKN Sequence homology: Cow: 85%; Dog: 92%; Guinea Pig: 85%; Horse: 92%; Human: 100%; Mouse: 92%; Pig: 92%; Rabbit: 92%; Rat: 92%; Yeast: 91%.
MORF4L2 is a protein coding gene. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when directly recruited to sites of DNA damage. Also component of the MSIN3A complex which acts to repress transcription by deacetylation of nucleosomal histones.
Specifications
MORF4L2 | |
Polyclonal | |
Unconjugated | |
MORF4L2 | |
2410017O14Rik; Kiaa0026; Liver regeneration-related protein LRRG00119; LRRG00119; mKIAA0026; Morf4l2; MORFL2; MORF-related gene X; MORF-related gene X protein; mortality factor 4 like 2; mortality factor 4-like protein 2; MRGX; MSL3-2 protein; OTTHUMP00000023756; protein MSL3-2; RP5-1055C14.2; Sid 393; Sid393; Sid393p; transcription factor-like protein Mrgx | |
Rabbit | |
Protein A | |
RUO | |
9643 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q15014 | |
MORF4L2 | |
Synthetic peptide directed towards the N-terminal of human MORF4L2 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction