Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MORN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA564450
Description
Immunogen sequence: SGVTYYGLWI NGHPAEQATR IVILGPEVME VAQGSPFSVN VQLLQDHGEI AKSESGRVLQ ISAGVRYVQL SAYSEVNFFK VDRDNQETLI QTPF Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 72%.
The function of this protein remains unknown.
Specifications
MORN1 | |
Polyclonal | |
Unconjugated | |
MORN1 | |
MORN repeat containing 1; MORN repeat-containing protein 1; MORN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79906 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T089 | |
MORN1 | |
Recombinant protein corresponding to Human MORN1. Recombinant protein control fragment (Product #RP-103984). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction