Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110700
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110700 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110700 Supplier Invitrogen™ Supplier No. PA5110700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YYEGAFVDGE ITGEGRRHWA WSGDTFSGQF VLGEPQGYGV MEYKAGGCYE GEVSHGMREG HGFLVDRDGQ VYQGSFHDNK RHGPGQMLFQ NGDKYDGDWV RDRRQGHGVL RCADGSTYKG QWHSDVFSGL GS Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 85%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORN1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MORN1
Gene Accession No. Q5T089
Gene Alias MORN repeat containing 1; MORN repeat-containing protein 1; MORN1
Gene Symbols MORN1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human MORN1. Recombinant protein control fragment (Product #RP-88838).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.