Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569947
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569947 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569947 Supplier Invitrogen™ Supplier No. PA569947
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

MORN4 gene ontology annotations related to this gene include response to axon injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORN4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MORN4
Gene Accession No. Q6PGF2
Gene Alias 44050 protein; bA548K23.4; C10orf83; C26H10orf83; chromosome 10 open reading frame 83; MORN repeat containing 4; MORN repeat-containing protein 4; MORN4; neuroprotective protein 4; protein 44050; Retinophilin; retinophilin homolog; RGD1307336; rtp
Gene Symbols MORN4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MORN4 (aa 96-145).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 226123
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.