Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562670
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562670 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562670 Supplier Invitrogen™ Supplier No. PA562670
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FNGVGVFIRY DNMTFEGEFK NGRVDGFGLL TFPDGSHGIP RNEGLFENNK LLRREKCSAI VQRAQSASKS ARNL Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.

MORN4 gene ontology annotations related to this gene include response to axon injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORN4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MORN4
Gene Accession No. Q8NDC4
Gene Alias 44050 protein; bA548K23.4; C10orf83; C26H10orf83; chromosome 10 open reading frame 83; MORN repeat containing 4; MORN repeat-containing protein 4; MORN4; neuroprotective protein 4; protein 44050; Retinophilin; retinophilin homolog; RGD1307336; rtp
Gene Symbols MORN4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MORN4. Recombinant protein control fragment (Product #RP-97169).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118812
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.